accessibility menu, dialog, popup

UserName

OOOHOOHOHH2ONa

CAS RN 6131-98-2

IUPAC Name: sodium 3-carboxy-2,3-dihydroxypropanoate hydrate
Synonyms: sodium 3-carboxy-2,3-dihydroxypropanoate hydrate
Molecular Weight (g/mol): 190.08
Molecular Formula: C4H7NaO7
InChi Key: LLVQEXSQFBTIRD-UHFFFAOYNA-M
SMILES: O.[Na+].OC(C(O)C([O-])=O)C(O)=O
Grade 
  • (2)
  • (1)
  • (4)
Molecular Weight (g/mol) 
  • (6)
  • (1)
Physical Form 
  • (2)
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
Percent Purity 
  • (4)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (6)
  • (1)
  • (6)
  • (2)
  • (1)
  • (4)
  • (6)
  • (1)
  • (2)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (4)
  • (2)