missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Bitartrate Monohydrate (Crystalline/Certified), Fisher Chemical
Supplier: Thermo Fisher Scientific S245500
Specifications
| Sodium Bitartrate Monohydrate | |
| White | |
| C4H7NaO7 | |
| MFCD00150038 | |
| sodium bitartrate monohydrate, sodium hydrogen tartrate monohydrate, sodium bitartrate,monohydrate, tartaric acid monosodium salt, sodium 3-carboxy-2,3-dihydroxypropanoate hydrate, l-+-tartaric acid monosodium salt, sodium bitartrate, monohydrate,, sodium bitartrate monohydrate, bioultra t, sodium 3-carboxy-2,3-dihydroxypropanoate-water 1/1/1, sodium bitartrate monohydrate, purum p.a t | |
| LLVQEXSQFBTIRD-UHFFFAOYSA-M | |
| sodium;2,3,4-trihydroxy-4-oxobutanoate;hydrate | |
| 23678966 | |
| Certified |
| 6131-98-2 | |
| Poly Bottle | |
| NaHC4H4O6·H2O | |
| 500 g | |
| 0.01% max. Insoluble in NH4OH | |
| C(C(C(=O)[O-])O)(C(=O)O)O.O.[Na+] | |
| 190.083 | |
| 190.09 | |
| Powder or Solid |
Chemical Identifiers
| 6131-98-2 | |
| 190.083 | |
| LLVQEXSQFBTIRD-UHFFFAOYSA-M | |
| 23678966 | |
| C(C(C(=O)[O-])O)(C(=O)O)O.O.[Na+] |
| C4H7NaO7 | |
| MFCD00150038 | |
| sodium bitartrate monohydrate, sodium hydrogen tartrate monohydrate, sodium bitartrate,monohydrate, tartaric acid monosodium salt, sodium 3-carboxy-2,3-dihydroxypropanoate hydrate, l-+-tartaric acid monosodium salt, sodium bitartrate, monohydrate,, sodium bitartrate monohydrate, bioultra t, sodium 3-carboxy-2,3-dihydroxypropanoate-water 1/1/1, sodium bitartrate monohydrate, purum p.a t | |
| sodium;2,3,4-trihydroxy-4-oxobutanoate;hydrate |
Safety and Handling