missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bisulfite (Powder/Certified ACS), Fisher Chemical™

Mixture of NaHSO3 and Na2S2O5

Supplier:  Thermo Fisher Scientific S6543

 View more versions of this product

Catalog No. S654-3



Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sodium Bisulfite
White
4 to 5
3 kg
MFCD00003530
DWAQJAXMDSEUJJ-UHFFFAOYSA-L
sodium bisulfite
23665763
≥58.5%
Pass Test
1.480g/cm³
150°C
7631-90-5,7681-57-4,7757-82-6
Powder/Solid
NaHSO3
sodium bisulfite, sodium hydrogen sulfite, sodium bisulphite, sodium hydrogensulfite, sodium sulfhydrate, monosodium sulfite, uantax sbs, sulfurous acid, monosodium salt, caswell no. 750, hydrogen sodium sulfite
[Na+].[O-]S([O-])=O
104.05
CHEBI:26709
Certified ACS
Poly Bottle

Chemical Identifiers

7631-90-5
104.05
DWAQJAXMDSEUJJ-UHFFFAOYSA-L
23665763
sodium bisulfite
NaHSO3
MFCD00003530
sodium bisulfite, sodium hydrogen sulfite, sodium bisulphite, sodium hydrogensulfite, sodium sulfhydrate, monosodium sulfite, uantax sbs, sulfurous acid, monosodium salt, caswell no. 750, hydrogen sodium sulfite
CHEBI:26709
[Na+].[O-]S([O-])=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

SDS Documents

Documents

Promotions

Promotions