missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bisulfite (Powder/Certified ACS), Fisher Chemical™

Catalog No. p-33650
Change view
Click to view available options
Quantity:
500 g
3 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S654500 500 g Poly Bottle
S6543 3 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S654500 Supplier Thermo Fisher Scientific Supplier No. S654 500
Edge

Mixture of NaHSO3 and Na2S2O5

Chemical Identifiers

CAS 7631-90-5 , 7681-57-4 , 7757-82-6
Molecular Formula NaHSO3
Molecular Weight (g/mol) 104.05
MDL Number MFCD00003530
InChI Key DWAQJAXMDSEUJJ-UHFFFAOYSA-L
Synonym sodium bisulfite, sodium hydrogen sulfite, sodium bisulphite, sodium hydrogensulfite, sodium sulfhydrate, monosodium sulfite, uantax sbs, sulfurous acid, monosodium salt, caswell no. 750, hydrogen sodium sulfite
PubChem CID 23665763
ChEBI CHEBI:26709
IUPAC Name sodium bisulfite
SMILES [Na+].[O-]S([O-])=O

Specifications

Melting Point 150°C
Color White
pH 4 to 5
Physical Form Powder/Solid
Quantity 500 g
Percent Purity ≥58.5%
Grade Certified ACS
Identification Pass Test
Packaging Poly Bottle
Density 1.480g/cm³
Chemical Name or Material Sodium Bisulfite
Show More Show Less