missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bisulfite, 10% (w/v), Ricca Chemical

Catalog No. p-7239360
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
714916 500 mL Natural Poly Bottle
714932 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 714916 Supplier Ricca Chemical Company Supplier No. 714916

APHA for Manganese Testing

Chemical Identifiers

CAS 7631-90-5
Molecular Formula NaO3S
Molecular Weight (g/mol) 103.05
InChI Key DWAQJAXMDSEUJJ-UHFFFAOYSA-L
IUPAC Name sodium sulfite
SMILES [Na+].[O-]S([O-])=O

Specifications

Percent Purity 10%
Grade Laboratory
Packaging Natural Poly Bottle
pH Acidic
Quantity 500 mL
Chemical Name or Material Sodium Bisulfite