missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bisulfate Monohydrate (Crystalline/Certified), Fisher Chemical™

Catalog No. p-33641
Change view
Click to view available options
Quantity:
500 g
3 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S240500 500 g Poly Bottle
S2403 3 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S240500 Supplier Thermo Fisher Scientific Supplier No. S240 500
Only null left

Chemical Identifiers

CAS 10034-88-5
Molecular Formula H3NaO5S
Molecular Weight (g/mol) 138.07
MDL Number MFCD00149210
InChI Key JXHZRQHZVYDRGX-UHFFFAOYSA-M
Synonym sodium bisulfate monohydrate, sodium hydrogen sulfate monohydrate, unii-3kls9y77yj, sulfuric acid, monosodium salt, hydrate, 3kls9y77yj, sodium hydrogensulfate hydrate, sulfuric acid, monosodium salt, monohydrate, sodium hydrate hydrogen sulfate, sodium hydrogen sulfate-1-hydrate, acmc-20ajox
PubChem CID 23673662
IUPAC Name sodium hydrate hydrogen sulfate
SMILES O.[Na+].OS([O-])(=O)=O

Specifications

Melting Point 58°C
Color White
pH 1
Physical Form Solid
Quantity 500 g
Formula Weight 138.07
Grade Certified
Identification Pass Test
Packaging Poly Bottle
Decomposition Information 430°C
Density 2.12g/cm³
Chemical Name or Material Sodium Bisulfate Monohydrate
Show More Show Less
Safety handling Check Alert storage code Red
DOTInformation DOT Class 8, : Corrosive