missing translation for 'onlineSavingsMsg'
Learn More

Sodium bisulfate monohydrate, 97%, extra pure, reagent, crystals

Catalog No. p-41771
Change view
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Packaging:
Glass bottle
Plastic drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC419435000 500 g Glass bottle
AC419430025 2.5 kg Glass bottle
AC419430100 10 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC419435000 Supplier Thermo Scientific Chemicals Supplier No. 419435000
Only null left

CAS: 10034-88-5 | H3NaO5S | 138.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10034-88-5
Molecular Formula H3NaO5S
Molecular Weight (g/mol) 138.07
MDL Number MFCD00149210
InChI Key JXHZRQHZVYDRGX-UHFFFAOYSA-M
Synonym sodium bisulfate monohydrate, sodium hydrogen sulfate monohydrate, unii-3kls9y77yj, sulfuric acid, monosodium salt, hydrate, 3kls9y77yj, sodium hydrogensulfate hydrate, sulfuric acid, monosodium salt, monohydrate, sodium hydrate hydrogen sulfate, sodium hydrogen sulfate-1-hydrate, acmc-20ajox
PubChem CID 23673662
IUPAC Name sodium;hydrogen sulfate;hydrate
SMILES O.[Na+].OS([O-])(=O)=O

Specifications

Melting Point 58.0°C
Color White
Physical Form Adhering Crystals
Quantity 500 g
Assay Percent Range 97%
Linear Formula NaHSO4·H2O
Merck Index 15,8722
Solubility Information Solubility in water: 670g/L. Other solubilities: decomposes by alcohol into sodium sulfate and, h2so4
Formula Weight 138.08
Percent Purity 97%
Grade Extra Pure
Packaging Glass bottle
Chemical Name or Material Sodium bisulfate monohydrate, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Precautionary Statement GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
RTECSNumber VZ1870000

RUO – Research Use Only