missing translation for 'onlineSavingsMsg'
Learn More

Sodium bismuthate(V), 85%

Catalog No. p-4509868
Change view
Click to view available options
Quantity:
25 g
250 g
1 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC205210250 25 g Glass bottle
AC205212500 250 g Glass bottle
AC205210010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC205210250 Supplier Thermo Scientific Chemicals Supplier No. 205210250
Only null left

CAS: 12232-99-4 | BiNaO3 | 279.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12232-99-4
Molecular Formula BiNaO3
Molecular Weight (g/mol) 279.96
InChI Key PNYYBUOBTVHFDN-UHFFFAOYSA-N
Synonym sodium bismuthate, bismuth sodium oxide, sodium bismuthate v, bismuth sodium trioxide, bismuthate bio31-, sodium, sodium oxido dioxo bismuth, sodium bismuth oxide, bio3.na, bismuthate bio31#-, sodium
PubChem CID 4063671
IUPAC Name sodium;oxido(dioxo)bismuth
SMILES [O-][Bi](=O)=O.[Na+]

Specifications

Color Yellow to Brown
Physical Form Powder
Quantity 25 g
Assay Percent Range 80% min. (Iodometry)
Linear Formula NaBiO3
Merck Index 15,8721
Solubility Information Solubility in water: insoluble. Other solubilities: decomposes in hot water and acids
Formula Weight 279.96
Percent Purity 85%
Packaging Glass bottle
Chemical Name or Material Sodium bismuthate(V)
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Do not eat,drink or smoke when using this product.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
If skin irritation occurs: Get medical advice/a
EINECSNumber 235-455-6

RUO – Research Use Only