accessibility menu, dialog, popup

UserName

NaOBiOO

CAS RN 12232-99-4

IUPAC Name: sodium bismuthanoylolate
Synonyms: sodium bismuthate
Molecular Weight (g/mol): 279.97
Molecular Formula: BiNaO3
InChi Key: PNYYBUOBTVHFDN-UHFFFAOYSA-N
SMILES: [Na+].[O-][Bi](=O)=O
Quantity 
  • (1)
  • (4)
  • (5)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (6)
  • (3)
Molecular Weight (g/mol) 
  • (5)
  • (5)
Grade 
  • (5)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (10)
  • (1)
  • (4)
  • (5)
  • (1)
  • (2)
  • (2)
  • (6)
  • (3)
  • (5)
  • (5)
  • (5)
  • (2)
  • (5)