missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bismuthate, Powder, ACS, 80%, Spectrum™ Chemical

Catalog No. 18613305
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613305 25 g Poly Bottle
18613306 100 g Poly Bottle
18613307 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18613305 Supplier Spectrum Chemical Mfg Cor Supplier No. S116525GM

Chemical Identifiers

CAS 12232-99-4

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 80%
Grade ACS
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.