missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bis(nonafluorobutanesulfonyl)imide 97.0+%, TCI America™

Catalog No. O04925G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O04925G 5 g
O04921G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O04925G Supplier TCI America Supplier No. O04925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 129135-86-0
Molecular Formula C8F18NNaO4S2
Molecular Weight (g/mol) 603.172
InChI Key NOIRUZKPVZNREY-UHFFFAOYSA-N
Synonym Bis(nonafluorobutanesulfonyl)imide Sodium Salt
PubChem CID 10930074
IUPAC Name sodium;bis(1,1,2,2,3,3,4,4,4-nonafluorobutylsulfonyl)azanide
SMILES C(C(C(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(C(F)(F)F)(F)F.[Na+]

Specifications

Color White
Quantity 5 g
UN Number 1759
Formula Weight 603.17
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Bis(nonafluorobutanesulfonyl)imide