accessibility menu, dialog, popup

UserName

FFSOONSOOFFFFFFFFFFFFFFFFNa

CAS RN 129135-86-0

IUPAC Name: sodium 1,1,1,2,2,3,3,4,4-nonafluoro-4-{[(1,1,2,2,3,3,4,4,4-nonafluorobutanesulfonyl)azanidyl]sulfonyl}butane
Synonyms: sodium 1,1,1,2,2,3,3,4,4-nonafluoro-4-[(1,1,2,2,3,3,4,4,4-nonafluorobutanesulfonyl)azanidylsulfonyl]butane
Molecular Weight (g/mol): 603.17
Molecular Formula: C8F18NNaO4S2
InChi Key: NOIRUZKPVZNREY-UHFFFAOYSA-N
SMILES: [Na+].FC(F)(F)C(F)(F)C(F)(F)C(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results