missing translation for 'onlineSavingsMsg'
Learn More

Sodium bis(2-methoxyethoxy)aluminum hydride, 70 wt% solution in toluene (ca. 3.5M), AcroSeal™

CAS: 22722-98-1 | C6H16AlNaO4 | 202.16 g/mol

$114.14 - $114.14

Chemical Identifiers

CAS 22722-98-1
Molecular Formula C6H16AlNaO4
Molecular Weight (g/mol) 202.16
MDL Number MFCD00011631
Synonym Sodium dihydro-bis-(2-methoxyethoxy)aluminate in toluene
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC430911000
SDS View Documents
Thermo Scientific Chemicals
430911000
100 mL AcroSeal™ Glass bottle
Each for $114.14
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
Specifications

Chemical Identifiers

22722-98-1
202.16
Sodium dihydro-bis-(2-methoxyethoxy)aluminate in toluene
C6H16AlNaO4
MFCD00011631

Specifications

110.0°C
22722-98-1 , 108-88-3
100 mL
[(CH3OCH2CH2O)2AlH2]Na
4°C
Sodium dihydro-bis-(2-methoxyethoxy)aluminate in toluene
202.16
68.5 to 71.5%
1.036
Sodium bis(2-methoxyethoxy)Aluminum hydride
Amber to Colorless
Viscous Liquid
C6H16AlNaO4
MFCD00011631
15,8702
Solubility in water: reacts. Other solubilities: soluble in aromatic hydrocarbons and ethers
202.16
AcroSeal™ Glass bottle
1.0360g/mL
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause drowsiness or dizziness.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
May be fata

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 245-178-2

SDS
Documents

Documents

RUO – Research Use Only