missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bicarbonate USP/FCC Powder, Macron Fine Chemicals™

macron

$6556.22 - $6556.22

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
Synonym sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
PubChem CID 516892
ChEBI CHEBI:32139
IUPAC Name sodium hydrogen carbonate
SMILES [Na+].OC([O-])=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
MK7396500
View Documents
Avantor Macron
739604
500 g POLYSTORMOR
Case of 12 Each for $6,556.22
Only null left
 
Description

Description

Specifications

Chemical Identifiers

144-55-8
84.01
UIIMBOGNXHQVGW-UHFFFAOYSA-M
516892
sodium hydrogen carbonate
CHNaO3
MFCD00003528
sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
CHEBI:32139
[Na+].OC([O-])=O

Specifications

144-55-8
500 g
MFCD00003528
UIIMBOGNXHQVGW-UHFFFAOYSA-M
sodium hydrogen carbonate
516892
84.01
POLYSTORMOR
Powder
CHNaO3
sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
[Na+].OC([O-])=O
84.01
CHEBI:32139
FCC/USP
Sodium Bicarbonate
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'