missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium bicarbonate/Sodium carbonate, IC eluent, Concentrate (20x) for Metrosep A Supp 10, MilliporeSigma™ Supelco™
This eluent concentrate is application tested. It is certified in accordance with ISO Guide 31. All details about exact content, uncertainty, traceability and expiry date are described in the certificate.
$274.07 - $274.07
Specifications
| CAS | 497-19-8 |
|---|---|
| Concentration or Composition (by Analyte or Components) | 100 mM Na2 CO3 aq. soln. |