missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium bicarbonate/Sodium carbonate, IC eluent, Concentrate (20x) for Metrosep A Supp 10, MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 497-19-8 |
| Quantity | 1 L |
| Concentration or Composition (by Analyte or Components) | 100 mM Na2 CO3 aq. soln. |