missing translation for 'onlineSavingsMsg'
Learn More

Sodium bicarbonate, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., USP, E500, 99.0-100.5%, powder, Honeywell Fluka™

Catalog No. 60044800
Change view
Click to view available options
Quantity:
1 kg
5 kg
4 x 5 kg
6 x 1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60044800 6 x 1 kg
6001988 1 kg
6001989 5 kg
60044799 4 x 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60044800 Supplier Honeywell Chemicals Supplier No. 134336X1KG
Only null left

Puriss., meets analytical specification of BP, FCC, Ph. Eur., USP, E500, 99.0-100.5%, powder

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
Synonym sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
PubChem CID 516892
ChEBI CHEBI:32139
IUPAC Name sodium;hydrogen carbonate
SMILES [Na+].OC([O-])=O

Specifications

Quantity 6 x 1 kg
Linear Formula NaHCO3
UN Number NONH for all modes of transport
Formula Weight 84.01
Percent Purity 99.0 to 100.5%
Grade puriss.
Chemical Name or Material Sodium Bicarbonate, Meets analytical specification of Ph. Eur.,BP, USP, FCC
EINECSNumber 205-633-8
RTECSNumber VZ0950000