missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Bicarbonate, Powder, U.S.P. - F.C.C., J.T. Baker™
Specifications
Specifications
| Physical Form | Powder |
| Quantity | 500 g |
| Formula Weight | 84.01 |
| Percent Purity | 100.5% |
| Grade | FCC/USP |
| Packaging | Wide Mouth Amber Glass Bottle |
| Specific Gravity | 1L = 2.20kg |
| Chemical Name or Material | Sodium Bicarbonate |