Learn More
Sodium Bicarbonate (Powder/Certified ACS), Fisher Chemical™
Supplier: Fisher Chemical S23350
Specifications
| Sodium Bicarbonate | |
| 144-55-8 | |
| 8.3 | |
| 50 kg | |
| CHNaO3 | |
| sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut | |
| [Na+].OC([O-])=O | |
| 84.01 | |
| CHEBI:32139 | |
| 99.7 to 100.3% | |
| Pass Test | |
| >50°C |
| 270°C | |
| White | |
| Powder/Solid | |
| 99.7 to 100.3 % | |
| MFCD00003528 | |
| UIIMBOGNXHQVGW-UHFFFAOYSA-M | |
| sodium hydrogen carbonate | |
| 516892 | |
| 84.01 | |
| Certified ACS | |
| Fiber Drum |
Chemical Identifiers
| 144-55-8 | |
| 84.01 | |
| UIIMBOGNXHQVGW-UHFFFAOYSA-M | |
| 516892 | |
| sodium hydrogen carbonate |
| CHNaO3 | |
| MFCD00003528 | |
| sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut | |
| CHEBI:32139 | |
| [Na+].OC([O-])=O |
Safety and Handling
WARNING!
Emergency Overview
Irritating to eyes and skin. May cause adverse kidney effects. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.
NFPA
Health:2
Flammability:0
Instability:1