missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Sodium Bicarbonate, Powder

Catalog No. MT61065RR
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MT61065RR 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MT61065RR Supplier Corning™ Supplier No. 61065RR

Part of buffering system commonly used to maintain physiological pH 7.2 to 7.4 in culture environment

Cellgro incorporates many principles developed by the pharmaceutical industry including the use of WFI quality water, raw material testing, in-process monitoring, and finished product testing.

  • Molecular weight: 84.01

Specifications

Content And Storage 15° to 30°C
Form Powder
Product Type Sodium Bicarbonate
Sterility Sterile-filtered
Format Plastic Bottle
pH 6.8 to 7.4
Quantity 1 kg