missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bicarbonate (Fine White Powder), Fisher BioReagents™

Catalog No. BP328500
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP328500 500 g Poly Bottle
BP3281 1 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP328500 Supplier Fisher BioReagents Supplier No. BP328 500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Edge

CAS: 144-55-8 | CHNaO3 | 84.01 g/mol

Sodium Bicarbonate (Fine White Powder)

This sodium salt is used in buffers for molecular biology and cell culture applications.

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
Synonym sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
PubChem CID 516892
ChEBI CHEBI:32139
IUPAC Name sodium hydrogen carbonate
SMILES [Na+].OC([O-])=O

Specifications

Melting Point 270°C
Color White
pH 8.3
Physical Form Powder/Solid
Quantity 500 g
Formula Weight 84.01
Percent Purity 99.7 to 100.3%
Identification Pass Test
Packaging Poly Bottle
Chemical Name or Material Sodium Bicarbonate
Safety handling Check Alert storage code Red
Recommended Storage RT