missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Bicarbonate (Fine White Powder), Fisher BioReagents™
Description
Sodium Bicarbonate (Fine White Powder)
This sodium salt is used in buffers for molecular biology and cell culture applications.
Specifications
Specifications
| Melting Point | 270°C |
| Color | White |
| pH | 8.3 |
| Physical Form | Powder/Solid |
| Quantity | 1 kg |
| Identification | Pass Test |
| Packaging | Poly Bottle |
| Formula Weight | 84.01 |
| Percent Purity | 99.7 to 100.3% |
| Grade | Ungraded |
| Show More |
Safety and Handling
| Recommended Storage | RT |