missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bicarbonate, ACS Grade, 99.7 to 100.3%, LabChem™

Catalog No. LC229431
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC229431 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC229431 Supplier LabChem Supplier No. LC229431

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
Synonym sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
PubChem CID 516892
ChEBI CHEBI:32139
IUPAC Name sodium hydrogen carbonate
SMILES [Na+].OC([O-])=O

Specifications

Melting Point 270°C
Color White
CAS Max % 100
Physical Form Powder
Quantity 500 g
Linear Formula NaHCO3
Solubility Information Soluble in water
Formula Weight 84.01
Percent Purity 99.7 to 100.3%
Grade ACS
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 2.159g/cm3
Decomposition Information Carbon monoxide; Carbon dioxide
Density 2.159g/cm3
Chemical Name or Material Sodium Bicarbonate
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Causes eye irritation.
Precautionary Statement GHS P Statement
Wash exposed skin thoroughly after handling.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
EINECSNumber 205-633-8
Recommended Storage Room Temperature