missing translation for 'onlineSavingsMsg'
Learn More

Sodium bicarbonate, 99.7+%, ACS reagent

Catalog No. p-4229027
Change view
Click to view available options
Quantity:
25 g
1 kg
5 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC424270250 25 g Plastic bottle
AC424270010 1 kg Plastic bottle
AC424270050 5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC424270250 Supplier Thermo Scientific Chemicals Supplier No. 424270250
Only null left

CAS: 144-55-8 | CHNaO3 | 84.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
Synonym sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
PubChem CID 516892
ChEBI CHEBI:32139
SMILES [Na+].OC([O-])=O

Specifications

Melting Point 270.0°C
Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.7+%
Linear Formula NaHCO3
Fieser 03,260; 05,595; 11,476; 17,180
Merck Index 15,8719
Solubility Information Solubility in water: 9 g/100mL (20°C). Other solubilities: 16.4 g/100mL water (60°C), insoluble in alcohol
IUPAC Name sodium;hydrogen carbonate
Formula Weight 84.01
Percent Purity ≥99.7%
Grade ACS Reagent
Packaging Plastic bottle
Chemical Name or Material Sodium bicarbonate
Show More Show Less
EINECSNumber 205-633-8

RUO – Research Use Only