missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Sodium bicarbonate, 99.5%, for biochemistry
CAS: 144-55-8 | CHNaO3 | 84.01 g/mol
Supplier: Thermo Scientific Chemicals 447102500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological bufferSpecifications
| Sodium bicarbonate | |
| 144-55-8 | |
| CHNaO3 | |
| MFCD00003528 | |
| sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut | |
| UIIMBOGNXHQVGW-UHFFFAOYSA-M | |
| sodium;hydrogen carbonate | |
| 516892 | |
| 84.01 | |
| Biochemical |
| 270.0°C | |
| 250 g | |
| NaHCO3 | |
| 158719 | |
| Clear colorless | |
| [Na+].OC([O-])=O | |
| 84.01 | |
| CHEBI:32139 | |
| 99.5% | |
| Plastic Bottle |
Chemical Identifiers
| 144-55-8 | |
| 84.01 | |
| UIIMBOGNXHQVGW-UHFFFAOYSA-M | |
| 516892 | |
| sodium;hydrogen carbonate |
| CHNaO3 | |
| MFCD00003528 | |
| sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut | |
| CHEBI:32139 | |
| [Na+].OC([O-])=O |
Safety and Handling
EINECSNumber : 205-633-8
RUO – Research Use Only