missing translation for 'onlineSavingsMsg'
Learn More

Sodium bicarbonate, 99.5%, for analysis

CAS: 144-55-8 | CHNaO3 | 84.01 g/mol

Supplier:  Thermo Scientific Chemicals 217120100

 View more versions of this product

Catalog No. AC217120100

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
Specifications

Specifications

Sodium bicarbonate
144-55-8
Fine Crystalline Powder
99.50%
NaHCO3
03,260; 05,595; 11,476; 17,180
sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
UIIMBOGNXHQVGW-UHFFFAOYSA-M
sodium hydrogen carbonate
516892
84.01
Analytical
270.0°C
White
10 kg
CHNaO3
MFCD00003528
15, 8719
Solubility in water: 9g/100mL (20°C). Other solubilities: 16.4g/100mL water (60°C),insoluble in alcohol
[Na+].OC([O-])=O
84.01
CHEBI:32139
99.5%
Plastic drum

Chemical Identifiers

144-55-8
84.01
UIIMBOGNXHQVGW-UHFFFAOYSA-M
516892
[Na+].OC([O-])=O
CHNaO3
MFCD00003528
sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
CHEBI:32139
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 205-633-8

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only