Learn More
Sodium Benzoate, Powder, NF, 99-101%, Spectrum™ Chemical
SO120, 532-32-1, C7H5O2Na
Supplier:  Spectrum Chemical Mfg Cor SO12025KG
 View more versions of this product
Catalog No. 18606320
Only null left
Explore available promotions
Description
Description
Spectrum™ Chemical Sodium Benzoate, Powder, NF is a tablet and capsule lubricant used in the pharmaceutical formulations. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
532-32-1
Poly Pail
MFCD00012463
WXMKPNITSTVMEF-UHFFFAOYSA-M
sodium benzoate
99.0 to 101%
1
C7H5NaO2
2.5 kg
[Na+].[O-]C(=O)C1=CC=CC=C1
144.11
NF
Chemical Identifiers
532-32-1
144.11
WXMKPNITSTVMEF-UHFFFAOYSA-M
[Na+].[O-]C(=O)C1=CC=CC=C1
C7H5NaO2
MFCD00012463
sodium benzoate
Product Suggestions
Product Suggestions