missing translation for 'onlineSavingsMsg'
Learn More

Sodium Benzoate, Powder, NF, 99-101%, Spectrum™ Chemical

Catalog No. p-7158183
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Catalog No. Quantity Packaging
18606319 500 g Poly Bottle
18606320 2.5 kg Poly Pail
18606321 12 kg Poly Pail
18606322 50 kg Fiber Drum
4 options

Catalog No. 18606319

Supplier: Spectrum Chemical Mfg Cor SO120500GM

Only null left

SO120, 532-32-1, C7H5O2Na

Spectrum™ Chemical Sodium Benzoate, Powder, NF is a tablet and capsule lubricant used in the pharmaceutical formulations. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 532-32-1
Molecular Formula C7H5NaO2
Molecular Weight (g/mol) 144.11
MDL Number MFCD00012463
InChI Key WXMKPNITSTVMEF-UHFFFAOYSA-M
IUPAC Name sodium benzoate
SMILES [Na+].[O-]C(=O)C1=CC=CC=C1

Specifications

CAS Max % 1
Packaging Poly Bottle
Quantity 500 g
Percent Purity 99.0 to 101%
Grade NF