missing translation for 'onlineSavingsMsg'
Learn More

Medisca Sodium Benzoate, NF

Catalog No. 14185658
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14185658 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14185658 Supplier Medisca Supplier No. 055108
Get 1 Extra Chemical When You Buy 4
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sodium Benzoate NF is a white, odorless or practically odorless, granular or crystalline powder. Hygroscopic. Alternative name(s): Sodium benzenecarboxylate

Sodium Benzoate NF is a white, odorless or practically odorless, granular or crystalline powder. Hygroscopic.

Alternative name(s): Sodium benzenecarboxylate

Specifications

Chemical Name or Material Sodium Benzoate, NF
Packaging Jar, 1000mL, White, HDPE, Round, 89-400, TE Pano Neck
CAS 532-32-1
Type Drug Standard
Molecular Weight (g/mol) 144.1
Quantity 500 g
Percent Purity 99 to 101 %
Grade NF