missing translation for 'onlineSavingsMsg'
Learn More

Sodium Azide, Crystalline, Laboratory, Fisher Chemical™

Supplier:  Thermo Fisher Scientific S227I100

 View more versions of this product

Catalog No. S227I-100

Only null left
Explore available promotions

Description

Description

  • Source of azide anion for organic chemical synthesis
  • Azide anions are strong nucleophiles and azide functionalized molecules can undergo multiple reactions
  • Handle with caution
  • Storage recommendations should be followed
  • For research use only
Specifications

Specifications

300°C
275°C
White
Powder/Solid
≥99 %
NaN3
sodium azide, azide, sodium, natriumazid, nemazyd, smite, kazoe, azoture de sodium, sodium azide na n3, hydrazoic acid, sodium salt, natriumazid german
[N-]=[N+]=[N-].[Na+]
65.011
CHEBI:278547
≥99%
Pass test
1.850g/cm³
Sodium Azide
26628-22-8
10
100 g
N3Na
MFCD00003536
PXIPVTKHYLBLMZ-UHFFFAOYSA-N
sodium;azide
33557
65.01
Laboratory
Poly Bottle

Chemical Identifiers

26628-22-8
65.011
PXIPVTKHYLBLMZ-UHFFFAOYSA-N
33557
sodium;azide
N3Na
MFCD00003536
sodium azide, azide, sodium, natriumazid, nemazyd, smite, kazoe, azoture de sodium, sodium azide na n3, hydrazoic acid, sodium salt, natriumazid german
CHEBI:278547
[N-]=[N+]=[N-].[Na+]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Emergency Overview
Very toxic if swallowed. Very toxic in contact with skin. May be fatal if absorbed through skin or swallowed. Toxic by inhalation. May cause pulmonary edema. Irritating to eyes, respiratory syste

Signal Word: DANGER!

DOTInformation : DOT Class 6.1, : Poison

SDS Documents

Documents

Promotions

Promotions