missing translation for 'onlineSavingsMsg'
Learn More

Sodium Azide, Crystalline, Laboratory, Fisher Chemical™

Catalog No. S227I25
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S227I25 25 g Poly Bottle
S227I100 100 g Poly Bottle
S227I500 500 g Poly Bottle
S227I1 1 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S227I25 Supplier Thermo Fisher Scientific Supplier No. S227I 25
Get Up to $1,800 in Chemicals with Purchase
View all promotions
  • Source of azide anion for organic chemical synthesis
  • Azide anions are strong nucleophiles and azide functionalized molecules can undergo multiple reactions
  • Handle with caution
  • Storage recommendations should be followed
  • For research use only

Chemical Identifiers

CAS 26628-22-8
Molecular Formula N3Na
Molecular Weight (g/mol) 65.011
MDL Number MFCD00003536
InChI Key PXIPVTKHYLBLMZ-UHFFFAOYSA-N
Synonym sodium azide, azide, sodium, natriumazid, nemazyd, smite, kazoe, azoture de sodium, sodium azide na n3, hydrazoic acid, sodium salt, natriumazid german
PubChem CID 33557
ChEBI CHEBI:278547
IUPAC Name sodium;azide
SMILES [N-]=[N+]=[N-].[Na+]

Specifications

Boiling Point 300°C
Melting Point 275°C
Color White
pH 10
Physical Form Powder/Solid
Quantity 25 g
Assay Percent Range ≥99 %
Linear Formula NaN3
Formula Weight 65.01
Percent Purity ≥99%
Grade Laboratory
Identification Pass test
Packaging Poly Bottle
Density 1.850g/cm³
Chemical Name or Material Sodium Azide
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Category Signal Word: DANGER!
Precautionary Statement Emergency Overview
Very toxic if swallowed. Very toxic in contact with skin. May be fatal if absorbed through skin or swallowed. Toxic by inhalation. May cause pulmonary edema. Irritating to eyes, respiratory syste
DOTInformation DOT Class 6.1, : Poison