missing translation for 'onlineSavingsMsg'
Learn More

Sodium Azide, Granular, 99%, Spectrum™ Chemical

Catalog No. 18613276
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
12 kg
2.5 kg
25 g
500 g
Packaging:
Amber Glass Bottle
Poly Pail
Catalog No. Quantity Packaging
18613276 12 kg Poly Pail
18613274 25 g Amber Glass Bottle
18613275 100 g Amber Glass Bottle
18613277 500 g Amber Glass Bottle
18613279 2.5 kg Amber Glass Bottle
5 options

Catalog No. 18613276

Supplier: Spectrum Chemical Mfg Cor SO11012KG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 26628-22-8
Molecular Formula N3Na
Molecular Weight (g/mol) 65.01
InChI Key PXIPVTKHYLBLMZ-UHFFFAOYSA-N
IUPAC Name sodium azide
SMILES [Na+].[N-]=[N+]=[N-]

Specifications

CAS Max % 1
Quantity 12 kg
Percent Purity 99%
Grade Reagent
Loss on Drying 0.001
Packaging Poly Pail

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.