missing translation for 'onlineSavingsMsg'
Learn More

Sodium Azide, Granular, 99%, Spectrum™ Chemical

Catalog No. 18613274
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
12 kg
Packaging:
Amber Glass Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613274 25 g Amber Glass Bottle
18613275 100 g Amber Glass Bottle
18613277 500 g Amber Glass Bottle
18613279 2.5 kg Amber Glass Bottle
18613276 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18613274 Supplier Spectrum Chemical Mfg Cor Supplier No. SO11025GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 26628-22-8
Molecular Formula N3Na
Molecular Weight (g/mol) 65.01
InChI Key PXIPVTKHYLBLMZ-UHFFFAOYSA-N
IUPAC Name sodium azide
SMILES [Na+].[N-]=[N+]=[N-]

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 99%
Grade Reagent
Loss on Drying 0.001
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.