missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Azide, ACS, 99%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor S166625KG
Specifications
| 26628-22-8 | |
| 2.5 kg | |
| PXIPVTKHYLBLMZ-UHFFFAOYSA-N | |
| sodium azide | |
| 99% | |
| 0.001 |
| 1 | |
| N3Na | |
| [Na+].[N-]=[N+]=[N-] | |
| 65.01 | |
| ACS | |
| Amber Glass Bottle |
Chemical Identifiers
| 26628-22-8 | |
| 65.01 | |
| sodium azide |
| N3Na | |
| PXIPVTKHYLBLMZ-UHFFFAOYSA-N | |
| [Na+].[N-]=[N+]=[N-] |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.