missing translation for 'onlineSavingsMsg'
Learn More

Sodium Azide, ACS, 99%, Spectrum™ Chemical

Catalog No. 18613280
Change view
Click to view available options
Quantity:
100 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613280 100 g Amber Glass Bottle
18613282 2.5 kg Amber Glass Bottle
18613281 12 kg Poly Pail
18613284 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18613280 Supplier Spectrum Chemical Mfg Cor Supplier No. S1666100GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 26628-22-8
Molecular Formula N3Na
Molecular Weight (g/mol) 65.01
InChI Key PXIPVTKHYLBLMZ-UHFFFAOYSA-N
IUPAC Name sodium azide
SMILES [Na+].[N-]=[N+]=[N-]

Specifications

CAS Max % 1
Quantity 100 g
Percent Purity 99%
Grade ACS
Loss on Drying 0.001
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.