missing translation for 'onlineSavingsMsg'
Learn More

Sodium Azide, ≥99%

Catalog No. p-7082757
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ2161022 100 g Plastic bottle
AAJ2161036 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ2161022 Supplier Thermo Fisher Scientific Supplier No. J2161022

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 26628-22-8 | N3Na | 65.011 g/mol

Chemical Identifiers

CAS 26628-22-8
Molecular Formula N3Na
Molecular Weight (g/mol) 65.011
MDL Number MFCD00003536
InChI Key PXIPVTKHYLBLMZ-UHFFFAOYSA-N
Synonym sodium azide, azide, sodium, natriumazid, nemazyd, smite, kazoe, azoture de sodium, sodium azide na n3, hydrazoic acid, sodium salt, natriumazid german
PubChem CID 33557
ChEBI CHEBI:278547
IUPAC Name sodium;azide
SMILES [N-]=[N+]=[N-].[Na+]

Specifications

Melting Point ∼275°C (decomposition)
Physical Form Crystalline Powder
Quantity 100 g
UN Number UN1687
Formula Weight 65.01
Percent Purity ≥99%
Packaging Plastic bottle
Specific Gravity 1.846
Density 1.846
Chemical Name or Material Sodium Azide
Hazard Category H300+H310+H330-H373
Precautionary Statement P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P314-P330-P361-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 247-852-1
RTECSNumber VY8050000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only