missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sodium azide, 99+%, for biochemistry

Sodium azide, CAS # 26628-22-8, is the sodium salt of hydrogen azide (hydrazoic acid). This compound shows antibacterial and mutagenic activities. It also can inhibit the mitochondrial respiratory chain. | CAS: 26628-22-8 | N3Na | 65.01 g/mol

Supplier:  Thermo Scientific Chemicals 447810250

 View more versions of this product

Catalog No. AC447810250


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Information

• Sodium azide is a precursor for preparing inorganic azides such as lead azide and silver azide
• One mechanism of action associated with this compound can be related to the inhibition of mitochondrial ATPase, that consequently suppresses oxidative phosphorylation

Applications

• Sodium azide is an inorganic compound with antibacterial and mutagenic activities mainly used in biological samples
• Sodium azide inhibits cytochrome oxidase in gram-negative bacteria. However, its antibacterial capacity is more limited in some gram-positive bacteria (streptococci, pneumococci, lactobacilli), presenting intrinsic resistance
• Sodium azide is used for the introduction of an azide group to prepare a desired compound
• It is useful in biochemical approaches as a probe reagent and a preservative
• This compound can be used to synthetize blue fluorescent copolymers and metal phosphonates

Specifications

Specifications

300.0°C
275.0°C
25 g
N3Na
15,8717
Solubility in water: 420g/L (17°C). Other solubilities: soluble in liquid ammonia, slightly soluble in alcohol, insoluble in diethyl ether
[N-]=[N+]=[N-].[Na+]
65.01
CHEBI:278547
≥99%
Plastic bottle
1.8500g/mL
Sodium azide
26628-22-8
99% min. (Argentometry)
NaN3
sodium azide, azide, sodium, natriumazid, nemazyd, smite, kazoe, azoture de sodium, sodium azide na n3, hydrazoic acid, sodium salt, natriumazid german
PXIPVTKHYLBLMZ-UHFFFAOYSA-N
sodium;azide
33557
65.01
Biochemical
1.85

Chemical Identifiers

26628-22-8
65.01
sodium azide, azide, sodium, natriumazid, nemazyd, smite, kazoe, azoture de sodium, sodium azide na n3, hydrazoic acid, sodium salt, natriumazid german
CHEBI:278547
[N-]=[N+]=[N-].[Na+]
N3Na
PXIPVTKHYLBLMZ-UHFFFAOYSA-N
33557
sodium;azide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Rinse mouth.
Wear protective gloves/protective clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.

GHS Signal Word: Danger

EINECSNumber : 247-852-1

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only