missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sodium azide, 99+%, for biochemistry

Catalog No. AC447811000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC447811000 100 g
AC447810250 25 g
AC447815000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC447811000 Supplier Thermo Scientific Chemicals Supplier No. 447811000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sodium azide, CAS # 26628-22-8, is the sodium salt of hydrogen azide (hydrazoic acid). This compound shows antibacterial and mutagenic activities. It also can inhibit the mitochondrial respiratory chain. ∣ CAS: 26628-22-8 ∣ N3Na ∣ 65.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Information

  • Sodium azide is a precursor for preparing inorganic azides such as lead azide and silver azide
  • One mechanism of action associated with this compound can be related to the inhibition of mitochondrial ATPase, that consequently suppresses oxidative phosphorylation

Applications

  • Sodium azide is an inorganic compound with antibacterial and mutagenic activities mainly used in biological samples
  • Sodium azide inhibits cytochrome oxidase in gram-negative bacteria. However, its antibacterial capacity is more limited in some gram-positive bacteria (streptococci, pneumococci, lactobacilli), presenting intrinsic resistance
  • Sodium azide is used for the introduction of an azide group to prepare a desired compound
  • It is useful in biochemical approaches as a probe reagent and a preservative
  • This compound can be used to synthetize blue fluorescent copolymers and metal phosphonates

Chemical Identifiers

CAS 26628-22-8
Molecular Formula N3Na
Molecular Weight (g/mol) 65.01
InChI Key PXIPVTKHYLBLMZ-UHFFFAOYSA-N
Synonym sodium azide, azide, sodium, natriumazid, nemazyd, smite, kazoe, azoture de sodium, sodium azide na n3, hydrazoic acid, sodium salt, natriumazid german
PubChem CID 33557
ChEBI CHEBI:278547
IUPAC Name sodium;azide
SMILES [N-]=[N+]=[N-].[Na+]

Specifications

Boiling Point 300.0°C
Melting Point 275.0°C
Physical Form Powder
Quantity 100 g
Packaging Plastic bottle
Specific Gravity 1.85
Assay Percent Range 99% min. (Argentometry)
Density 1.8500g/mL
Linear Formula NaN3
Merck Index 15,8717
Solubility Information Solubility in water: 420g/L (17°C). Other solubilities: soluble in liquid ammonia, slightly soluble in alcohol, insoluble in diethyl ether
Formula Weight 65.01
Percent Purity ≥99%
Grade Biochemical
Chemical Name or Material Sodium azide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Rinse mouth.
Wear protective gloves/protective clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
EINECSNumber 247-852-1

RUO – Research Use Only