missing translation for 'onlineSavingsMsg'
Learn More

Sodium aurothiomalate(I), 99.9% (metals basis)

Catalog No. AA3974003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3974003 1 g
AA3974006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3974003 Supplier Thermo Scientific Chemicals Supplier No. 03974003

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 12244-57-4 | C4H5AuNa2O5S | 408.09 g/mol

Sodium aurothiomalate(I) is useful for high mobility group box chromosomal protein 1 western blotting.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12244-57-4
Molecular Formula C4H5AuNa2O5S
Molecular Weight (g/mol) 408.09
MDL Number MFCD00064304,MFCD00064304
InChI Key YLQOAPBVYJCTPW-UHFFFAOYNA-K
Synonym gold sodium thiomalate
PubChem CID 133108869
IUPAC Name gold;sodium;2-sulfanylbutanedioic acid
SMILES O.[Na+].[Na+].[Au+].[O-]C(=O)CC([S-])C([O-])=O

Specifications

Odor Sulphurous
Quantity 1 g
Merck Index 14,4518
Solubility Information Very soluble in water; Practically insoluble in alcohol, ether
Formula Weight 390.08
Percent Purity 99.9%
Physical Form Powder
Assay (metals basis)
Chemical Name or Material Sodium aurothiomalate(I)
Hazard Category H302+H332-H317
Hazard Statement GHS H Statement
H301-H332-H317
Toxic if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P270-P271-P272-P280g-P301+P312-P302+P352-P304+P340-P312-P330-P333+P313-P363-P501c
EINECSNumber 235-479-7
RTECSNumber MD5435000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only