missing translation for 'onlineSavingsMsg'
Learn More

Sodium ascorbate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012968
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012968 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012968 Supplier Merck Emd Millipore Supplier No. PHR12791G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sodium ascorbate, also known as E301, is a sodium salt of ascorbic acid. It is generally used as an antioxidant in pharmaceutical formulations and food products.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 134-03-2
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C6H7 NaO6
MDL Number MFCD00082340
Melting Point 220°C
Synonym (+)-Sodium L-ascorbate; L(+)-Ascorbic acid sodium salt; Vitamin C sodium salt
Type Certified Reference Material
Quantity 1 g
Formula Weight 198.11
Grade Certified Reference Material
Show More Show Less