missing translation for 'onlineSavingsMsg'
Learn More

Sodium antimonyl L-tartrate, 98+%

Catalog No. p-7020671
Change view
Click to view available options
Quantity:
50 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1270718 50 g
AAA127070B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1270718 Supplier Thermo Scientific Chemicals Supplier No. A1270718
Only null left

CAS: 34521-09-0 | C8H4Na2O12Sb2

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 34521-09-0
Molecular Formula C8H4Na2O12Sb2
MDL Number MFCD00065402

Specifications

Quantity 50 g
UN Number UN1549
Merck Index 14,702
Formula Weight 581.6
Percent Purity ≥98%
Chemical Name or Material Sodium antimonyl L-tartrate
Hazard Category H302+H332
Hazard Statement GHS H Statement
H331-H302
Toxic if inhaled.
Harmful if swallowed.
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ANTIMONY COMPOUNDS, INORGANIC, SOLID, N.O.S.
EINECSNumber 252-070-9
RTECSNumber CC8050000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only