accessibility menu, dialog, popup

UserName

NaNaSbOOSbOOOOOOOOOO

CAS RN 34521-09-0

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 581.61
Molecular Formula: C8H4Na2O12Sb2
InChi Key: UZVPADYOEPQJSP-UHFFFAOYNA-J
SMILES: [Na+].[Na+].[Sb+3].[Sb+3].[O-]C(C([O-])C([O-])=O)C([O-])=O.[O-]C(C([O-])C([O-])=O)C([O-])=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results