missing translation for 'onlineSavingsMsg'
Learn More

Sodium Anthraquinone-2-sulfonate Monohydrate 98.0+%, TCI America™

Catalog No. p-7116939
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A050825G 25 g
A0508500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A050825G Supplier TCI America Supplier No. A050825G

Chemical Identifiers

CAS 153277-35-1
Molecular Formula C14H7NaO5S
Molecular Weight (g/mol) 310.25
MDL Number MFCD00001229
InChI Key GGCZERPQGJTIQP-UHFFFAOYSA-M
Synonym sodium 9,10-dioxo-9,10-dihydroanthracene-2-sulfonate hydrate, anthraquinone-2-sulfonic acid sodium salt monohydrate, sodium anthraquinone-2-sulfonate monohydrate, sodium 9,10-dioxoanthracene-2-sulfonate hydrate, 9,10-anthraquinone-2-sulfonic acid sodium salt hydrate, anthraquinone-2-sulfonic acid, sodium salt, monohydrate, 9,10-dihydro-9,10-dioxo-2-anthracenesulfonic acid sodium salt, acmc-209bkc, c14h8o5s.na.h2o, anthraquinone-2-sulfonic acid sodium salt hydrate
PubChem CID 23666331
IUPAC Name sodium 9,10-dioxo-9,10-dihydroanthracene-2-sulfonate
SMILES [Na+].[O-]S(=O)(=O)C1=CC=C2C(=O)C3=CC=CC=C3C(=O)C2=C1

Specifications

Color Yellow
Quantity 25 g
Formula Weight 310.25
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Anthraquinone-2-sulfonate Monohydrate
EINECSNumber (4)-0736
RTECSNumber CB1095550
TSCA Yes