accessibility menu, dialog, popup

UserName

OSOOOONa

CAS RN 153277-35-1

IUPAC Name: sodium 9,10-dioxo-9,10-dihydroanthracene-2-sulfonate
Synonyms: sodium 9,10-dioxoanthracene-2-sulfonate
Molecular Weight (g/mol): 310.25
Molecular Formula: C14H7NaO5S
InChi Key: GGCZERPQGJTIQP-UHFFFAOYSA-M
SMILES: [Na+].[O-]S(=O)(=O)C1=CC=C2C(=O)C3=CC=CC=C3C(=O)C2=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)