missing translation for 'onlineSavingsMsg'
Learn More

Sodium Anthraquinone-1-sulfonate 98.0+%, TCI America™

Catalog No. A142825G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A142825G 25 g
A1428250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A142825G Supplier TCI America Supplier No. A142825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 128-56-3
Molecular Formula C14H7NaO5S
Molecular Weight (g/mol) 310.255
MDL Number MFCD00037145
InChI Key SDKPSXWGRWWLKR-UHFFFAOYSA-M
Synonym Anthraquinone-1-sulfonic Acid Sodium Salt
PubChem CID 517266
IUPAC Name sodium;9,10-dioxoanthracene-1-sulfonate
SMILES C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=CC=C3)S(=O)(=O)[O-].[Na+]

Specifications

Color Yellow
Quantity 25 g
Formula Weight 310.25
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Anthraquinone-1-sulfonate
EINECSNumber (4)-0736
RTECSNumber CB1095540
TSCA Yes