accessibility menu, dialog, popup

UserName

CAS RN 128-56-3

IUPAC Name: sodium 9,10-dioxo-9,10-dihydroanthracene-1-sulfonate
Synonyms: sodium 9,10-dioxoanthracene-1-sulfonate
Molecular Weight (g/mol): 310.25
Molecular Formula: C14H7NaO5S
InChi Key: SDKPSXWGRWWLKR-UHFFFAOYSA-M
SMILES: [Na+].[O-]S(=O)(=O)C1=CC=CC2=C1C(=O)C1=CC=CC=C1C2=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results