missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Ammonium Phosphate, Saturated, Ricca Chemical
Supplier: Ricca Chemical Company R71370001A
Specifications
| Sodium Ammonium Phosphate | |
| 7783-13-3 , 7732-18-5 | |
| H13NNaO8P | |
| IQTQISLCLWJRPM-UHFFFAOYSA-M | |
| azanium;sodium;hydrogen phosphate;tetrahydrate | |
| 56924540 | |
| Natural Poly Bottle |
| Saturated | |
| 1 L | |
| unii-k78u8wlv4h, ammonium sodium phosphate tetrahydrate, k78u8wlv4h, phosphorsalz, acmc-20ajp4, ammonium sodium phosphate tetrahydrate mi, sodium monoammonium monohydrogen phosphate tetrahydrate, phosphoric acid, monoammonium monosodium salt, tetrahydrate, salt of phosphorus microcosmic salt, sodium ammonium phosphate tetrahydrate | |
| [NH4+].O.O.O.O.OP(=O)([O-])[O-].[Na+] | |
| 209.067 | |
| Odorless |
Chemical Identifiers
| 7783-13-3 | |
| 209.067 | |
| unii-k78u8wlv4h, ammonium sodium phosphate tetrahydrate, k78u8wlv4h, phosphorsalz, acmc-20ajp4, ammonium sodium phosphate tetrahydrate mi, sodium monoammonium monohydrogen phosphate tetrahydrate, phosphoric acid, monoammonium monosodium salt, tetrahydrate, salt of phosphorus microcosmic salt, sodium ammonium phosphate tetrahydrate | |
| azanium;sodium;hydrogen phosphate;tetrahydrate |
| H13NNaO8P | |
| IQTQISLCLWJRPM-UHFFFAOYSA-M | |
| 56924540 | |
| [NH4+].O.O.O.O.OP(=O)([O-])[O-].[Na+] |
Safety and Handling
ShelfLife : 24 months