Learn More
Sodium Aluminate, Technical, Spectrum™ Chemical
S1120, 1302-42-7, Na2O·Al2O3·3H2O
Supplier:  Spectrum Chemical Mfg Cor S112012KG
 View more versions of this product
Catalog No. 18606293
Only null left
Explore available promotions
Description
Description
Spectrum™ Chemical Sodium Aluminate, Technical is an inorganic chemical that is used as in water softening systems, as a coagulant aid to improve flocculation, and for removing dissolved silica and phosphates. Technical grade products supplied by Spectrum are indicative of a grade suitable for general industrial use. These materials may or may not have a Certificate of Analysis available.
Specifications
Specifications
1302-42-7
12 kg
UN2812
[Na+].O=[Al-]=O
81.97
Poly Pail
1
AlNaO2
IYJYQHRNMMNLRH-UHFFFAOYSA-N
sodium dioxoalumanuide
Technical
Chemical Identifiers
1302-42-7
81.97
sodium dioxoalumanuide
AlNaO2
IYJYQHRNMMNLRH-UHFFFAOYSA-N
[Na+].O=[Al-]=O
Product Suggestions
Product Suggestions