missing translation for 'onlineSavingsMsg'
Learn More

Sodium Alginate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015829
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015829 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015829 Supplier Merck Emd Millipore Supplier No. PHR14711G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Sodium Alginate is the sodium salt of alginic acid, extracted from sea weeds. It is generally used as a stabilizer and thickener in the food industry.

Specifications

CAS 9005-38-3
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
MDL Number MFCD00081310
Synonym Alginic acid sodium salt; Algin; Sodium alginate
Type Certified Reference Material
Quantity 1 g
Grade Certified Reference Material