Learn More
Sodium Alginate, FCC, 90.8-106%, Spectrum™ Chemical
SO106, 9005-38-3, (C6H7O6Na)n
Supplier:  Spectrum Chemical Mfg Cor SO10612KG
 View more versions of this product
Catalog No. 18606285
Only null left
Explore available promotions
Description
Description
Spectrum™ Chemical Sodium Alginate, FCC is a gum that has no discernible flavor and is used in the food industry as an emulsifier and to increase viscosity. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
9005-38-3
(C6H7O7)A(C6H7O7)BNa
[Na+].O[C@H]1[C@@H](O)[C@H](O-*)[C@H](O[C@H]1O-*)C([O-])=O.O[C@H]1[C@@H](O)[C@H](O-*)[C@@H](O[C@H]1O-*)C([O-])=O
FCC
Poly Pail
1
12 kg
90.8 to 106%
0.15
Chemical Identifiers
9005-38-3
(C6H7O7)A(C6H7O7)BNa
[Na+].O[C@H]1[C@@H](O)[C@H](O-*)[C@H](O[C@H]1O-*)C([O-])=O.O[C@H]1[C@@H](O)[C@H](O-*)[C@@H](O[C@H]1O-*)C([O-])=O
Product Suggestions
Product Suggestions